Learn More
Biomatik Corporation Human CNN1 (Calponin-1) ELISA Kit
Supplier:  Biomatik Corporation EKF57425
Encompass_Preferred
Human CNN1 (Calponin-1) ELISA Kit for detecting CNN1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P51911. Target Synonyms: CNN1; Calponin-1; Calponin H1; Sm-Calp; SMCC; Basic calponin; calponin 1; basic; smooth muscle; Calponin H1; smooth muscle; calponins; basic. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5341