Learn More
Biomatik Corporation Human S100P (Protein S100-P) ELISA Kit
Supplier:  Biomatik Corporation EKF57436
Encompass_Preferred
Human S100P (Protein S100-P) ELISA Kit for detecting S100P in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P25815. Target Synonyms: S100P; Protein S100-P; MIG9; migRation-inducing gene 9; Protein S100-E; S100 calcium binding protein P; S100 calcium-binding protein Pprotein S100-P; S100E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5349