Learn More
Biomatik Corporation Human HIG2 (Hypoxia-inducible gene 2 protein) ELISA Kit
Supplier:  Biomatik Corporation EKF57456
Encompass_Preferred
Human HIG2 (Hypoxia-inducible gene 2 protein) ELISA Kit for detecting HIG2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9Y5L2. Target Synonyms: HIG2; C7orf68; FLJ21076; HIG-2; HIG2.. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5354