Learn More
Biomatik Corporation Human CASP7 (Caspase-7) ELISA Kit
Supplier:  Biomatik Corporation EKF57478
Encompass_Preferred
Human CASP7 (Caspase-7) ELISA Kit for detecting CASP7 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P55210. Target Synonyms: Human CASP7; Caspase-7; ICE-like apoptotic protease 3; ICE-LAP3; Apoptotic protease Mch-3; CMH-1; CASP-7; MCH3; CASP7; Mch3; apoptosis-related cysteine protease; Apoptotic protease Mch-3; CASP-7; caspase 7; apoptosis-related cysteine peptidase; ICE-like apoptotic protease 3; Lice2 alpha; beta; gamma; CASP7; MCH3; Apoptotic protease Mch-3; CMH-1; ICE-like apoptotic protease 3; ICE-LAP3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5367