Learn More
Biomatik Corporation Human KRT1 (KeRatin, type II cytoskeletal 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57482
Encompass_Preferred
Human KRT1 (KeRatin, type II cytoskeletal 1) ELISA Kit for detecting KRT1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04264. Target Synonyms: KRT1; Type-II keRatin Kb1; KeRatin-1; K1; Hair alpha protein; 67 kDa cytokeRatin; CytokeRatin-1; CK-1; KRTA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5371