Learn More
Biomatik Corporation Human IFNA1 (Interferon alpha-1/13) ELISA Kit
Supplier:  Biomatik Corporation EKF58781
Encompass_Preferred
Human IFNA1 (Interferon alpha-1/13) ELISA Kit for detecting IFNA1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01562. Target Synonyms: IFNA1; Interferon alpha-1; 13. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6047