Learn More
Biomatik Corporation Mouse MMP-8 (Matrix Metalloproteinase 8) ELISA Kit
Supplier:  Biomatik Corporation EKF58805
Encompass_Preferred
Mouse MMP-8 (Matrix Metalloproteinase 8) ELISA Kit for detecting MMP-8 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O70138. Target Synonyms: MMP-8; Matrix Metalloproteinase 8; Collagenase 2; CLG1HNC; neutrophil collagenase; PMNL collagenase; PMNL-CL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6068