Learn More
Biomatik Corporation Mouse Nts (Neurotensin/neuromedin N) ELISA Kit
Supplier:  Biomatik Corporation EKF58809
Encompass_Preferred
Mouse Nts (Neurotensin/neuromedin N) ELISA Kit for detecting Nts in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.563-100pg/ml. Sensitivity: 0.938pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: Q9D3P9. Target Synonyms: Nts; Neurotensin; neuromedin N. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6072