Learn More
Biomatik Corporation Human FcγR3A (Fc Fragment of IgG Low Affinity IIIa Receptor) ELISA Kit
Supplier:  Biomatik Corporation EKF58815
Encompass_Preferred
Human FcγR3A (Fc Fragment of IgG Low Affinity IIIa Receptor) ELISA Kit for detecting FcγR3A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.813-500ng/ml. Sensitivity: 4.688ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08637. Target Synonyms: FcγR3A; CD16a; FCGR3A; CD16a; CD16a antigen; CD16FCRIIIA; Fc fragment of IgG; low affinity IIIa; receptor; CD16a; Fc fragment of IgG; low affinity IIIa; receptor for; CD16; Fc gamma receptor III-A; FCG3; Fc-gamma receptor III-2; CD 16; Fc-gamma receptor IIIb; CD16; Fc-gamma RIII; Fc-gamma RIIIa; Fc-gamma RIII-alpha; FCGR3; FCGRIII; FcR-10; FCR-10; FcRIII; FCRIII; FcRIIIa; IGFR3FcgammaRIIIA; immunoglobulin G Fc receptor III; low affinity III; receptor for. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6076