Learn More
Biomatik Corporation Rat Ctss (Cathepsin S) ELISA Kit
Supplier:  Biomatik Corporation EKF58820
Encompass_Preferred
Rat Ctss (Cathepsin S) ELISA Kit for detecting Ctss in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q02765. Target Synonyms: Ctss; Cathepsin S. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6080