Learn More
Biomatik Corporation Rat sRANKL (Soluble Receptor Activator of Nuclear factor-kB Ligand) ELISA Kit
Supplier:  Biomatik Corporation EKF58824
Encompass_Preferred
Rat sRANKL (Soluble Receptor Activator of Nuclear factor-kB Ligand) ELISA Kit for detecting sRANKL in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9ESE2. Target Synonyms: sRANKL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6083