Learn More
Biomatik Corporation Rat TGFBI (Transforming Growth Factor Beta Induced Protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58826
Encompass_Preferred
Rat TGFBI (Transforming Growth Factor Beta Induced Protein) ELISA Kit for detecting TGFBI in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: D4A8G5. Target Synonyms: TGFBI; βIG-H3; Transforming growth factor-beta-induced protein ig-h3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6085