Learn More
Biomatik Corporation Human A1BG (Alpha-1B-glycoprotein) ELISA Kit
Supplier:  Biomatik Corporation EKF58829
Encompass_Preferred
Human A1BG (Alpha-1B-glycoprotein) ELISA Kit for detecting A1BG in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04217. Target Synonyms: α1BG; Alpha-1-B-Glycoprotein; A1BG; alpha 1B-Glycoprotein; A1B; ABG; GAB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6088