Learn More
Biomatik Corporation Human DHH (Desert hedgehog protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58833
Encompass_Preferred
Human DHH (Desert hedgehog protein) ELISA Kit for detecting DHH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O43323. Target Synonyms: DHH; Desert hedgehog protein; HHG-3; desert hedgehog; desert hedgehog homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6091