Learn More
Biomatik Corporation Human IFIT1 (Interferon-induced protein with tetRatricopeptide repeats 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58835
Encompass_Preferred
Human IFIT1 (Interferon-induced protein with tetRatricopeptide repeats 1) ELISA Kit for detecting IFIT1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P09914. Target Synonyms: IFIT1; Interferon-induced protein with tetRatricopeptide repeats 1; IFIT-1; Interferon-induced 56 kDa protein; IFI-56K; G10P1; IFI56; IFNAI1; ISG56; GARG-16; IFI-56; ISG56; RNM561. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6093