Learn More
Biomatik Corporation Human CD86 (T-lymphocyte activation antigen CD86) ELISA Kit
Supplier:  Biomatik Corporation EKF58855
Encompass_Preferred
Human CD86 (T-lymphocyte activation antigen CD86) ELISA Kit for detecting CD86 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P42081. Target Synonyms: B7-2; CD86; Activation B7-2 antigen; B70; B7-2 antigen; B-lymphocyte activation antigen B7-2; BU63; CD28 antigen ligand 2; CD28LG2; CD86 antigen; CD86 molecule; CTLA-4 counter-receptor B7.2; FUN-1; LAB72; T-lymphocyte activation antigen CD86. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6108