Learn More
Biomatik Corporation Mouse Serpina3n (Serine protease inhibitor A3N) ELISA Kit
Supplier:  Biomatik Corporation EKF58884
Encompass_Preferred
Mouse Serpina3n (Serine protease inhibitor A3N) ELISA Kit for detecting Serpina3n in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q91WP6. Target Synonyms: Serpina3n; Spi2; Serine protease inhibitor A3N; Serpin A3N. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6131