Learn More
Biomatik Corporation Rat TEK (Tyrosine Kinase, Endothelial) ELISA Kit
Supplier:  Biomatik Corporation EKF58892
Encompass_Preferred
Rat TEK (Tyrosine Kinase, Endothelial) ELISA Kit for detecting TEK in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: D3ZCD0. Target Synonyms: Tek; CD202b; TEK; angiopoietin-1 receptor; CD202B; CD202b antigen; hTIE2; p140 TEK; soluble TIE2 variant 1; soluble TIE2 variant 2; TEK tyrosine kinase; endothelial; TIE-2; TIE2CD202b; Tunica interna endothelial cell kinase; Tyrosine-protein kinase receptor TEK; Tyrosine-protein kinase receptor TIE-2; venous malformations; multiple cutaneous and mucosal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6137