Learn More
Biomatik Corporation Mouse CXCR3 (CXC-chemokine receptor 3) ELISA Kit
Supplier:  Biomatik Corporation EKF58904
Encompass_Preferred
Mouse CXCR3 (CXC-chemokine receptor 3) ELISA Kit for detecting CXCR3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O88410. Target Synonyms: CXCR3; CD183; GPR9; CD183 antigen; chemokine; C-X-C motif receptor 3; chemokine; C-X-C receptor 3; CKR-L2IP-10 receptor; CMKAR3; C-X-C chemokine receptor type 3; CXC-R3; CXCR-3; G protein-coupled receptor 9; CD182; Mig-R; Interferon-inducible protein 10 receptor; IP10 receptor; IP10-R; Mig receptor; MigR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6144