Learn More
Biomatik Corporation Mouse GC (Glucagon) ELISA Kit
Supplier:  Biomatik Corporation EKF58909
Encompass_Preferred
Mouse GC (Glucagon) ELISA Kit for detecting GC in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P55095. Target Synonyms: GCG; Glucagon; GLP1; GLP2; GRPP; glicentin-related polypeptide; glucagon-like peptide 1; glucagon-like peptide 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6148