Learn More
Biomatik Corporation Human ZG16 (Zymogen granule membrane protein 16) ELISA Kit
Supplier:  Biomatik Corporation EKF58925
Encompass_Preferred
Human ZG16 (Zymogen granule membrane protein 16) ELISA Kit for detecting ZG16 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O60844. Target Synonyms: ZG16; Zymogen granule membrane protein 16; Secretory lectin ZG16; Zymogen granule protein 16; hZG16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6160