Learn More
Biomatik Corporation Mouse WISP2 (WNT1-inducible-signaling pathway protein 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58934
Encompass_Preferred
Mouse WISP2 (WNT1-inducible-signaling pathway protein 2) ELISA Kit for detecting WISP2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9Z0G4. Target Synonyms: WISP2; Ccn5; Ctgfl; WISP-2; CTGF-L; CCN family member 5; Connective tissue growth factor-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6166