Learn More
Biomatik Corporation Mouse CT (Calcitonin) ELISA Kit
Supplier:  Biomatik Corporation EKF58950
Encompass_Preferred
Mouse CT (Calcitonin) ELISA Kit for detecting CT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P70160. Target Synonyms: CT; Calcitonin; Thyrocalcitonin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6178