Learn More
Biomatik Corporation Mouse Hsd17b13 (17-beta-hydroxysteroid dehydrogenase 13) ELISA Kit
Supplier:  Biomatik Corporation EKF58961
Encompass_Preferred
Mouse Hsd17b13 (17-beta-hydroxysteroid dehydrogenase 13) ELISA Kit for detecting Hsd17b13 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8VCR2. Target Synonyms: Hsd17b13; Alcohol dehydrogenase PAN1B-like; Short-chain dehydrogenase; reductase 9;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6186