Learn More
Biomatik Corporation Human CAMKK2 (Calcium/calmodulin-dependent protein kinase kinase 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58965
Encompass_Preferred
Human CAMKK2 (Calcium/calmodulin-dependent protein kinase kinase 2) ELISA Kit for detecting CAMKK2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q96RR4. Target Synonyms: CAMKK2; CaM-KK 2; CaM-kinase kinase 2; CaMKK 2; Calcium; calmodulin-dependent protein kinase kinase beta; CaM-KK beta; CaM-kinase kinase beta; CaMKK beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6189