Learn More
Biomatik Corporation Human TNC (Tenascin) ELISA Kit
Supplier:  Biomatik Corporation EKF58979
Encompass_Preferred
Human TNC (Tenascin) ELISA Kit for detecting TNC in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P24821. Target Synonyms: TNC; Tenascin C; Tenascin; JI; Hexabrachion; Glioma-associated-extracellular matrix antigen; GP 150-225; Neuronectin; Myotendinous antigen; Cytotactin; GMEM; HXB; Tenascin J1; Cytotactin; Glioma-associated-extracellular matrix antigen; Hexabrachion; hexabrachion; tenascin; HXBcytotactin; TN-C; TNGP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6200