Learn More
Biomatik Corporation Rat SIRT1 (NAD-dependent protein deacetylase sirtuin-1) ELISA Kit
Supplier:  Biomatik Corporation EKF58990
Encompass_Preferred
Rat SIRT1 (NAD-dependent protein deacetylase sirtuin-1) ELISA Kit for detecting SIRT1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: A0A0G2JZ79. Target Synonyms: SIRT1; SIR2alpha; SIR2L1; SIR2-like protein 1; EC 3.5.1; S. cerevisiae; homolog 1; sirtuin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6209