Learn More
Biomatik Corporation Human APH1A (Gamma-secretase subunit APH-1A) ELISA Kit
Supplier:  Biomatik Corporation EKF58994
Encompass_Preferred
Human APH1A (Gamma-secretase subunit APH-1A) ELISA Kit for detecting APH1A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q96BI3. Target Synonyms: APH1A; Gamma-secretase subunit APH-1A; APH-1; APH-1a; aph-1alpha; Aph-1alpha; CGI-78; PSF; anterior pharynx defective 1 homolog A; Presenilin-stabilization factor; Aph-1alpha; Presenilin-stabilization factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6213