Learn More
Biomatik Corporation Human GZMK (Granzyme K) ELISA Kit
Supplier:  Biomatik Corporation EKF58996
Encompass_Preferred
Human GZMK (Granzyme K) ELISA Kit for detecting GZMK in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P49863. Target Synonyms: GZMK; NK-tryptase-2; NK-Tryp-2; Fragmentin-3; Granzyme-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6215