Learn More
Biomatik Corporation Human LGALS1 (Galectin-1) ELISA Kit
Supplier:  Biomatik Corporation EKF59005
Encompass_Preferred
Human LGALS1 (Galectin-1) ELISA Kit for detecting LGALS1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P09382. Target Synonyms: GAL1; Galectin 1; LGALS1; BHL; Galaptin; GBP; L-14; 14 kDa laminin-binding protein; Gal-1; galectin 1; galectin-1; GBP; HBL; HLBP14; HPL; Lactose-binding lectin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6223