Learn More
Biomatik Corporation Human LAMC2 (Laminin subunit gamma-2) ELISA Kit
Supplier:  Biomatik Corporation EKF59024
Encompass_Preferred
Human LAMC2 (Laminin subunit gamma-2) ELISA Kit for detecting LAMC2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q13753. Target Synonyms: LAMC2; Laminin subunit gamma-2; Laminin B2t chain; Ladsin 140 kDa subunit; Kalinin; nicein; epiligrin 100 kDa subunit; Kalinin subunit gamma; Nicein subunit gamma; Large adhesive scatter factor 140 kDa subunit; Laminin-5 subunit gamma; Cell-scattering factor 140 kDa subunit; CSF 140 kDa subunit; Epiligrin subunit gamma; LAMB2T; LAMNB2; B2T; BM600; CSF; EBR2; EBR2A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6238