Learn More
Biomatik Corporation Human TLR2 (Toll-like receptor 2) ELISA Kit
Supplier:  Biomatik Corporation EKF59027
Encompass_Preferred
Human TLR2 (Toll-like receptor 2) ELISA Kit for detecting TLR2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O60603. Target Synonyms: TLR2; CD282; CD282 antigen; TIL4CD282; toll; interleukin 1 receptor-like 4; Toll; interleukin-1 receptor-like protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6241