Learn More
Biomatik Corporation Human LNPEP (Leucyl-cystinyl aminopeptidase) ELISA Kit
Supplier:  Biomatik Corporation EKF59047
Encompass_Preferred
Human LNPEP (Leucyl-cystinyl aminopeptidase) ELISA Kit for detecting LNPEP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9UIQ6. Target Synonyms: LNPEP; Leucyl-cystinyl aminopeptidase; IRAP; Otase; Oxytocinase; P-LAP; CAP; PLAP; Vasopressinase; angiotensin IV receptor; AT; 4 receptor; Cystinyl aminopeptidase; Insulin-regulated membrane aminopeptidase; Insulin-responsive aminopeptidase; Placental leucine aminopeptidase; insulin-regulated aminopeptidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6256