Learn More
Biomatik Corporation Human NOX1 (NADPH oxidase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57497
Encompass_Preferred
Human NOX1 (NADPH oxidase 1) ELISA Kit for detecting NOX1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9Y5S8. Target Synonyms: NOX1; NOH-1; Mitogenic oxidase 1; MOX-1; NADH; NADPH mitogenic oxidase subunit P65-MOX; Nicotinamide Adenine Dinucleotide Phosphate Oxidase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5376