Learn More
Biomatik Corporation Human 14-3-3 protein eta (YWHAH) ELISA Kit
Supplier:  Biomatik Corporation EKF57525
Encompass_Preferred
Human 14-3-3 protein eta (YWHAH) ELISA Kit for detecting 14-3-3 protein eta in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q04917. Target Synonyms: YWHA1; Protein AS1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5391