Learn More
Biomatik Corporation Human Ang I (Angiotensin-1) ELISA Kit
Supplier:  Biomatik Corporation EKF57534
Encompass_Preferred
Human Ang I (Angiotensin-1) ELISA Kit for detecting Ang I in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 75pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P01019. Target Synonyms: Ang I; Angiotensin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5396