Learn More
Biomatik Corporation Human DAT (Dopamine Transporter) ELISA Kit
Supplier:  Biomatik Corporation EKF57587
Encompass_Preferred
Human DAT (Dopamine Transporter) ELISA Kit for detecting DAT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q01959. Target Synonyms: DAT; Dopamine Transporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5414