Learn More
Biomatik Corporation Human Ep-CAM/CD326 (Epithelial Cell Adhesion Molecule) ELISA Kit
Supplier:  Biomatik Corporation EKF57596
Encompass_Preferred
Human Ep-CAM/CD326 (Epithelial Cell Adhesion Molecule) ELISA Kit for detecting Ep-CAM; CD326 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100iu/ml. Sensitivity: 0.938iu/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P16422. Target Synonyms: Ep-CAM; CD326; 17-1A; GA733-2; gp40; KS1; 4; M4S1; TACSTD1; TROP1; 17-1A; 323; A3; ACSTD1; antigen identified by monoclonal AUA1; CD326 antigen; Cell surface glycoprotein Trop-1; chromosome 4; surface marker; 35kD glycoprotein; DIAR5; EGP; EGP-2; EGP314; EGP40; EpCAM; Epithelial cell surface antigen; Epithelial glycoprotein; Epithelial glycoprotein 314; ESA; hEGP314; HNPCC8; KS 1; 4 antigen; KS1; 4; KSAHEA125; M1S2; M4S1Ly74; Major gastrointestinal tumor-associated protein GA733-2; MIC18MH99; MOC31. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5416