Learn More
Biomatik Corporation Human FGFR2 (Fibroblast Growth Factor Receptor 2) ELISA Kit
Supplier:  Biomatik Corporation EKF57607
Encompass_Preferred
Human FGFR2 (Fibroblast Growth Factor Receptor 2) ELISA Kit for detecting FGFR2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P21802. Target Synonyms: FGFR2; CD332; BFR-1; CD332 antigen; CFD1; craniofacial dysostosis 1; EC 2.7.10; EC 2.7.10.1; fibroblast growth factor receptor 2; FLJ98662; Jackson-Weiss syndrome; JWS; KeRatinocyte growth factor receptorreceptor like 14; KSAM; K-sam; soluble FGFR4 variant 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5420