Learn More
Biomatik Corporation Human GSP (Glycosylated Serum Protein) ELISA Kit
Supplier:  Biomatik Corporation EKF57629
Encompass_Preferred
Human GSP (Glycosylated Serum Protein) ELISA Kit for detecting GSP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000nmol/ml. Sensitivity: 46.875nmol/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02768. Target Synonyms: GSP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5428