Learn More
Biomatik Corporation Human LOX (Lysyl Oxidase) ELISA Kit
Supplier:  Biomatik Corporation EKF57648
Encompass_Preferred
Human LOX (Lysyl Oxidase) ELISA Kit for detecting LOX in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P28300. Target Synonyms: LOX; Protein-Lysine 6-Oxidase; MGC105112; Lysyl oxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5431