Learn More
Biomatik Corporation Human MT3 (Metallothionein 3) ELISA Kit
Supplier:  Biomatik Corporation EKF57657
Encompass_Preferred
Human MT3 (Metallothionein 3) ELISA Kit for detecting MT3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P25713. Target Synonyms: MT3; Metallothionein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5436