Learn More
Biomatik Corporation Human STAT1 (Signal Transducer and Activator of Transcription 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57723
Encompass_Preferred
Human STAT1 (Signal Transducer and Activator of Transcription 1) ELISA Kit for detecting STAT1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P42224. Target Synonyms: STAT1; CANDF7; ISGF-3; STAT91; DKFZp686B04100; ISGF-3; 91kD; signal transducer and activator of transcription 1; 91kDa; signal transducer and activator of transcription-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5459