Learn More
Biomatik Corporation Rat FE (Ferritin) ELISA Kit
Supplier:  Biomatik Corporation EKF57931
Encompass_Preferred
Rat FE (Ferritin) ELISA Kit for detecting FE in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.125-200pg/ml. Sensitivity: 1.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P19132; P02793. Target Synonyms: Ferritin; FE. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5471