Learn More
Biomatik Corporation Rat GA (Glycated Albumin) ELISA Kit
Supplier:  Biomatik Corporation EKF57972
Encompass_Preferred
Rat GA (Glycated Albumin) ELISA Kit for detecting GA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pmol/ml. Sensitivity: 9.375pmol/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02770. Target Synonyms: Glycated Albumin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5477