Learn More
Biomatik Corporation Human PON2 (Serum paraoxonase/arylesterase 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58002
Encompass_Preferred
Human PON2 (Serum paraoxonase/arylesterase 2) ELISA Kit for detecting PON2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q15165. Target Synonyms: PON2; Serum paraoxonase; arylesterase 2; A-esterase 2; Serum Aryldialkylphosphatase 2; A-esterase 2; Aromatic esterase 2; paraoxonase 2; paraoxonase nirs; PON 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5481