Learn More
Biomatik Corporation Mouse RAP1A (Ras-related protein Rap-1A) ELISA Kit
Supplier:  Biomatik Corporation EKF58032
Encompass_Preferred
Mouse RAP1A (Ras-related protein Rap-1A) ELISA Kit for detecting RAP1A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P62835. Target Synonyms: RAP1A; KREV1; Ras-related protein Krev-1; GTP-binding protein smg p21A; C21KG; G-22K. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5496