Learn More
Biomatik Corporation Rat ANP (Atrial Natriuretic Peptide) ELISA Kit
Supplier:  Biomatik Corporation EKF58043
Encompass_Preferred
Rat ANP (Atrial Natriuretic Peptide) ELISA Kit for detecting ANP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01161. Target Synonyms: ANP; NPPA; ANF; Cardionatrin; NPPA; PND; ANF; ANPatriopeptin; ATFB6; atrial natriuretic factor; Atrial natriuretic peptide; CDD-ANF; natriuretic peptide A; natriuretic peptide precursor A; PNDcardionatrin; prepronatriodilatin; Prepronatriodilatin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5503