Learn More
Biomatik Corporation Mouse Ctss (Cathepsin S) ELISA Kit
Supplier:  Biomatik Corporation EKF58077
Encompass_Preferred
Mouse Ctss (Cathepsin S) ELISA Kit for detecting Ctss in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O70370. Target Synonyms: Ctss; CTSS; cathepsin S; EC 3.4.22; EC 3.4.22.27; FLJ50259; MGC3886. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5524