Learn More
Biomatik Corporation Rat HA (Hyaluronic Acid) ELISA Kit
Supplier:  Biomatik Corporation EKF58080
Encompass_Preferred
Rat HA (Hyaluronic Acid) ELISA Kit for detecting HA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: Hyaluronic Acid. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5526